Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-nicotinamide adenine dinucleotide, reduced disodium salt | 606-68-8 | MFCD00036200 | 100.0% | 709.40 g·mol⁻¹ | C21H27N7Na2O14P2 | 10 MG
SDP

Catalog No. 5000297419
Encompass_Preferred

NADH disodium salt is the reduced form of nicotinamide adenine dinucleotide supplied as the sodium disalt for research and analytical applications; it functions as an electron donor in biochemical assays and metabolic studies.

  • High purity reagent suitable for analytical use.
  • Soluble in water and DMSO (reported up to 100 mg/mL).
  • Molecular weight 709.40 g·mol⁻¹; CAS number 606-68-8.
  • Storage: keep sealed, away from moisture; solid at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research sizes, including 10 MG packaging.

Catalog No. 50-002-97419 Supplier Medchemexpress LLC Supplier No. HYF0001R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NADH disodium salt is the reduced form of nicotinamide adenine dinucleotide supplied as the sodium disalt for research and analytical applications; it functions as an electron donor in biochemical assays and metabolic studies.

  • High purity reagent suitable for analytical use.
  • Soluble in water and DMSO (reported up to 100 mg/mL).
  • Molecular weight 709.40 g·mol⁻¹; CAS number 606-68-8.
  • Storage: keep sealed, away from moisture; solid at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research sizes, including 10 MG packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.