Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Nicotinamide adenine100MG
Catalog No. 502897305
Encompass
-Nicotinamide adenine dinucleotide (NAD) is an electron carrier and a cofactor. It is ubiquitously found in the living system. NAD contains an adenylic acid and a nicotinamide-5AE-ribonucleotide group linked together by a pyrophosphate moiety. In NAD complexes the enzyme-cofactor interactions are highly conserved.
Catalog No. 50-289-7305 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N1636100MG
-Nicotinamide adenine dinucleotide (NAD) is an electron carrier and a cofactor. It is ubiquitously found in the living system. NAD contains an adenylic acid and a nicotinamide-5AE-ribonucleotide group linked together by a pyrophosphate moiety. In NAD complexes the enzyme-cofactor interactions are highly conserved.