Learn More
Selleck Chemical LLC -Nicotinamide Mononucleotide 25mg 1094-61-7
SDP
Supplier:  Selleck Chemical LLC S525925MG
Encompass_Preferred
Nicotinamide mononucleotide (NMN), one of the key precursors of NAD+ and products of the NAMPT reaction , is converted to NAD+ by nicotinamide mononucleotide adenylyltransferase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8101