Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-phenylethyl beta-D-glucoside | 18997-54-1 | MFCD04972099 | ≥97.0% | 284.31 g·mol⁻¹ | C14H20O6 | 10 MG
SDP

Catalog No. 5000285244
Encompass_Preferred

Beta-phenylethyl beta-D-glucoside is a research-grade glycoside provided as a purified powder for biochemical and pharmacological studies. It is the β-anomer of phenylethyl glucoside (CAS 18997-54-1), with molecular formula C14H20O6 and molecular weight 284.31 g·mol⁻¹. Typical purity is ≥97.0% and the material is supplied in small milligram quantities for analytical and preparative use.

  • High purity (≥97.0%) suitable for analytical applications.
  • Powder form for accurate weighing and formulation.
  • Available in small milligram packages for method development.
  • Molecular formula and weight provided for calculations.
  • CAS number provided for unambiguous identification.
  • Datasheet and certificate of analysis available for quality verification.

Catalog No. 50-002-85244 Supplier Medchemexpress LLC Supplier No. HYW35714010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta-phenylethyl beta-D-glucoside is a research-grade glycoside provided as a purified powder for biochemical and pharmacological studies. It is the β-anomer of phenylethyl glucoside (CAS 18997-54-1), with molecular formula C14H20O6 and molecular weight 284.31 g·mol⁻¹. Typical purity is ≥97.0% and the material is supplied in small milligram quantities for analytical and preparative use.

  • High purity (≥97.0%) suitable for analytical applications.
  • Powder form for accurate weighing and formulation.
  • Available in small milligram packages for method development.
  • Molecular formula and weight provided for calculations.
  • CAS number provided for unambiguous identification.
  • Datasheet and certificate of analysis available for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.