Learn More
AdipoGen beta-Pix mouse pAb
SDP
Supplier:  AdipoGen AG25A0006C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the multiple splice variants of human, mouse and rat beta-Pix a, b, bL, c and d-isoforms. Cross-reacts with human and rat beta-Pix. Source Rabbit. Applications ELISA, ICC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. beta-PIX acts as a RAC1 guanine nucleotide exchange factor GEF and can induce membrane ruffling. It interacts with PAK kinases through the SH3 domain. It may function as a positive regulator of apoptosis and is involved in cell migration. It is expressed with high levels in the central nervous system.
Catalog No. 50-256-3305