Learn More
Medchemexpress LLC Beta-propiolactone | 57-57-8 | MFCD00005169 | 99.4% | 72.06 g/mol | C3H4O2 | 1 ML
SDP
Catalog No. 5000241941
Encompass_Preferred
Propiolactone (beta-propiolactone) is a small, reactive four-membered lactone (C3H4O2, 72.06 g/mol) provided as a 10 mM solution in DMSO for laboratory research. It is commonly used as a viral chemical inactivator and as a reactive reagent in biochemical and virology workflows. Handle with appropriate personal protective equipment and consult the SDS before use.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research applications.
  • Reactive lactone used for viral inactivation and chemical modification.
  • Stable when stored frozen; follow storage recommendations.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-41941 Supplier Medchemexpress LLC Supplier No. HY10793110MM 1ML
Propiolactone (beta-propiolactone) is a small, reactive four-membered lactone (C3H4O2, 72.06 g/mol) provided as a 10 mM solution in DMSO for laboratory research. It is commonly used as a viral chemical inactivator and as a reactive reagent in biochemical and virology workflows. Handle with appropriate personal protective equipment and consult the SDS before use.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research applications.
  • Reactive lactone used for viral inactivation and chemical modification.
  • Stable when stored frozen; follow storage recommendations.
  • For research use only; not for human or clinical use.