Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-propiolactone | 57-57-8 | MFCD00005169 | 99.4% | 72.06 g/mol | C3H4O2 | 250 MG
SDP

Catalog No. 5000241786
Encompass_Preferred

Propiolactone (beta-propiolactone) is a small, reactive lactone used primarily as a chemical viral inactivator and bacteriostatic agent in research and vaccine formulation. It is supplied as a high-purity reagent for laboratory applications that require reliable viral inactivation; the chemical is reactive and should be handled with appropriate safety precautions.

  • High purity (99.36%) suitable for research use.
  • Molecular weight 72.06 g/mol for analytical calculations.
  • Used as a chemical viral inactivator and bacteriostatic agent.
  • Available in multiple pack sizes, including 250 mg.
  • Chemical formula C3H4O2; synonyms include β-propiolactone and 2-oxetanone.

Catalog No. 50-002-41786 Supplier Medchemexpress LLC Supplier No. HY107931250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propiolactone (beta-propiolactone) is a small, reactive lactone used primarily as a chemical viral inactivator and bacteriostatic agent in research and vaccine formulation. It is supplied as a high-purity reagent for laboratory applications that require reliable viral inactivation; the chemical is reactive and should be handled with appropriate safety precautions.

  • High purity (99.36%) suitable for research use.
  • Molecular weight 72.06 g/mol for analytical calculations.
  • Used as a chemical viral inactivator and bacteriostatic agent.
  • Available in multiple pack sizes, including 250 mg.
  • Chemical formula C3H4O2; synonyms include β-propiolactone and 2-oxetanone.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.