Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-propiolactone | 57-57-8 | MFCD00005169 | 99.4% | 72.06 g/mol | C3H4O2 | 500 MG
SDP

Catalog No. 5000241942
Encompass_Preferred

Propiolactone (beta-propiolactone) is a small cyclic ester used in laboratory research as a chemical viral inactivator and analytical reagent. Supplied as a high-purity liquid, it is used for virus inactivation, preparation of analytical standards, and other controlled research applications and must be handled with appropriate safety precautions.

  • High purity (≈99.4%).
  • Molecular weight 72.06 g/mol.
  • Colorless to light yellow liquid with density ~1.1460 g/cm3.
  • Available in multiple pack sizes, including 500 mg.
  • Storage: pure form -20 °C; in solvent -80 °C (short term).
  • Used as a viral inactivator and analytical standard in research settings.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-41942 Supplier Medchemexpress LLC Supplier No. HY107931500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propiolactone (beta-propiolactone) is a small cyclic ester used in laboratory research as a chemical viral inactivator and analytical reagent. Supplied as a high-purity liquid, it is used for virus inactivation, preparation of analytical standards, and other controlled research applications and must be handled with appropriate safety precautions.

  • High purity (≈99.4%).
  • Molecular weight 72.06 g/mol.
  • Colorless to light yellow liquid with density ~1.1460 g/cm3.
  • Available in multiple pack sizes, including 500 mg.
  • Storage: pure form -20 °C; in solvent -80 °C (short term).
  • Used as a viral inactivator and analytical standard in research settings.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.