Learn More
Research Products International Corp Beta RackBox with Built-In Key Lock and Two 4-Way Flipper Racks, 3/8 Inches Thick, Clear Acrylic
SDP
Catalog No. 50550193
Encompass
Beta RackBox with built-in key lock, provides convenient access to tubes when working with radioactive samples. Removable end panel allows rack to slide in or out. Unique hinged lid can be positioned at any angle to serve as a frontal shield. Two 4-Way Flipper racks are included.
Constructed of heavy duty 3/8 inch thick clear acrylic for shielding 32P beta emissions. Safe for freezer storage to -70°C.
Catalog No. 50-550-193 Supplier Research Products International Corp Supplier No. 145756
Beta RackBox with built-in key lock, provides convenient access to tubes when working with radioactive samples. Removable end panel allows rack to slide in or out. Unique hinged lid can be positioned at any angle to serve as a frontal shield. Two 4-Way Flipper racks are included.
Constructed of heavy duty 3/8 inch thick clear acrylic for shielding 32P beta emissions. Safe for freezer storage to -70°C.