Learn More
AdipoGen beta-Rubromycin
SDP
Supplier:  AdipoGen BVT0251M001
Encompass_Preferred
Chemical. CAS 27267-70-5. Formula C27H20O12. MW 536.4. Isolated from Streptomyces sp. Antibiotic. HIV-1 reverse transcriptase inhibitor. Human telomerase inhibitor. Cytostatic against different tumor cell lines.
Catalog No. 50-256-2046