Learn More
Fisherbrand™ Beta Sample Box Interchangeable Racks
SCI_ED
Catalog No. S29141 Shop All Fisherbrand Products
Change view
Click to view available options
Tube Size:
2.5 mL
For Use With (Equipment):
For 2.5 mL tubes, 36-place box
Catalog No. Tube Size For Use With (Equipment)
S29141 2.5 mL For 2.5 mL tubes, 36-place box
1 options
Catalog No. S29141
For use with Beta Boxes.
  • 36-place box
  • Removable racks acco mmodate microcentrifuge tubes
  • Allow icing of samples if desired
  • Nonskid feet
  • With radiation label
Specifications
Includes Radiation label, non-skid feet
For Use With (Application) To accommodate microcentrifuge tubes
For Use With (Equipment) For 2.5 mL tubes, 36-place box
Product Type Interchangeable rack
Tube Size 2.5 mL