Learn More
CPC Scientific Z-Val-Leu-Leu-CHO 1MG
SDP
Catalog No. 502108742
Encompass
 SEQUENCE: Z-Val-Leu-Leu-CHOONE-LETTER SEQUENCE: ZVLL-CHOMOLECULAR FORMULA: C25H39N3O5MOLECULAR WEIGHT: 461.59STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [263563-09-3]SYNONYMS: -Secretase Inhibitor IIRESEARCH AREA: Alzheimer's DiseaseREFERENCES:G. Abbenante, et al., Biochem. Biophys. Res. Commun. 268, 133 (2000)SKU(s): AMYD-113A
Catalog No. 50-210-8742 Supplier CPC Scientific Supplier No. AMYD113A
 SEQUENCE: Z-Val-Leu-Leu-CHOONE-LETTER SEQUENCE: ZVLL-CHOMOLECULAR FORMULA: C25H39N3O5MOLECULAR WEIGHT: 461.59STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [263563-09-3]SYNONYMS: -Secretase Inhibitor IIRESEARCH AREA: Alzheimer's DiseaseREFERENCES:G. Abbenante, et al., Biochem. Biophys. Res. Commun. 268, 133 (2000)SKU(s): AMYD-113A