Learn More
Medchemexpress LLC Beta-secretase inhibitor IV | 797035-11-1 | 99.2% | 578.72 g·mol⁻¹ | C31H38N4O5S | 10 MG
SDP
Catalog No. 5000211381
Encompass_Preferred
A potent, cell-active small-molecule inhibitor of β-secretase (BACE-1) used in neuroscience research to reduce amyloid-β peptide production. The compound shows low-nanomolar enzymatic potency and demonstrable cellular activity, is supplied as a solid powder with high analytical purity, and is soluble in DMSO for in vitro work.
  • Potent BACE-1 inhibition with IC50 values around 15-16 nM.
  • Demonstrated cellular activity (EC50 ~43 nM) for reducing Aβ production.
  • High purity (approximately 99.2%) suitable for research applications.
  • Well-defined physicochemical properties: molecular weight ~578.72 g·mol⁻¹; formula C31H38N4O5S.
  • High solubility in DMSO (≈100 mg/mL) for stock preparation.
  • Stable as powder at -20 °C; storage conditions provided for solutions.
  • Available in small research pack sizes, including 10 mg, for laboratory use.
Catalog No. 50-002-11381 Supplier Medchemexpress LLC Supplier No. HY1013310MG
Add to Cart
Add to Cart
A potent, cell-active small-molecule inhibitor of β-secretase (BACE-1) used in neuroscience research to reduce amyloid-β peptide production. The compound shows low-nanomolar enzymatic potency and demonstrable cellular activity, is supplied as a solid powder with high analytical purity, and is soluble in DMSO for in vitro work.
  • Potent BACE-1 inhibition with IC50 values around 15-16 nM.
  • Demonstrated cellular activity (EC50 ~43 nM) for reducing Aβ production.
  • High purity (approximately 99.2%) suitable for research applications.
  • Well-defined physicochemical properties: molecular weight ~578.72 g·mol⁻¹; formula C31H38N4O5S.
  • High solubility in DMSO (≈100 mg/mL) for stock preparation.
  • Stable as powder at -20 °C; storage conditions provided for solutions.
  • Available in small research pack sizes, including 10 mg, for laboratory use.