Learn More
Medchemexpress LLC Beta-secretase inhibitor IV | 797035-11-1 | MFCD09037559 | 99.2% | 578.72 | C31H38N4O5S | 1 MG
SDP
Catalog No. 5000211383
Encompass_Preferred
β-Secretase Inhibitor IV is a potent, cell-permeable small-molecule inhibitor of BACE-1 used for in vitro studies of β-secretase activity and Aβ production. It exhibits low-nanomolar enzymatic potency and reduces Aβ peptide levels in cell-based assays.
Catalog No. 50-002-11383 Supplier Medchemexpress LLC Supplier No. HY101331MG
β-Secretase Inhibitor IV is a potent, cell-permeable small-molecule inhibitor of BACE-1 used for in vitro studies of β-secretase activity and Aβ production. It exhibits low-nanomolar enzymatic potency and reduces Aβ peptide levels in cell-based assays.