Learn More
CPC Scientific MCA-Glu-Val-Lys-Met-Asp-Ala-Glu-Phe-Lys(DNP)-NH2 (ammonium acetate salt) 1MG
SDP
Catalog No. 502108744
Encompass
ONE-LETTER SEQUENCE: Mca-EVKMDAEF-K(DNP)-NH2MOLECULAR FORMULA: C66H86N14O23S1MOLECULAR WEIGHT:1463.52STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: M. Farzan, et al., Proc. Natl. Acad. Sci. USA 97, 9712 (2000); T. Kowalewski, and D.M. Holtzman, Proc. Natl. Acad. Sci. USA 96, 3688 (1999); U.S. Ladror, et al., J. Protein. Chem. 13, 357 (1994)
Catalog No. 50-210-8744 Supplier CPC Scientific Supplier No. AMYD114B
ONE-LETTER SEQUENCE: Mca-EVKMDAEF-K(DNP)-NH2MOLECULAR FORMULA: C66H86N14O23S1MOLECULAR WEIGHT:1463.52STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: M. Farzan, et al., Proc. Natl. Acad. Sci. USA 97, 9712 (2000); T. Kowalewski, and D.M. Holtzman, Proc. Natl. Acad. Sci. USA 96, 3688 (1999); U.S. Ladror, et al., J. Protein. Chem. 13, 357 (1994)