Learn More
Research Products International Corp Multi-Comp Beta Shield, Four Compartment Shield
SDP
Catalog No. 50213044
Encompass
Accommodate stock solution vials containing 32P individual compartments limit exposure to one vial at a time. Supplied with inserts to accommodate smaller vials.
Catalog No. 50-213-044 Supplier Research Products International Corp Supplier No. 146514
Accommodate stock solution vials containing 32P individual compartments limit exposure to one vial at a time. Supplied with inserts to accommodate smaller vials.