Learn More
Medchemexpress LLC HY-N0171 5g , Beta-Sitosterol β-Sitosterol;22,23-Dihydrostigmasterol CAS:83-46-5 Purity:98%
SDP
Catalog No. 501492205
Encompass_Preferred
Medchemexpress, HY-N0171 5g Beta-Sitosterol β-Sitosterol;22,23-Dihydrostigmasterol CAS:83-46-5 Formula:C29H50O IC50: 99.99±1.86 μg/mL (PPL) Purity:98% Beta-Sitosterol weakly inhibits porcine pancreatic lipase (PPL) activity. Sitosterol is an important compound extracted from the leaves of Aloe vera . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2205 Supplier Medchemexpress LLC Supplier No. HYN01715G
Medchemexpress, HY-N0171 5g Beta-Sitosterol β-Sitosterol;22,23-Dihydrostigmasterol CAS:83-46-5 Formula:C29H50O IC50: 99.99±1.86 μg/mL (PPL) Purity:98% Beta-Sitosterol weakly inhibits porcine pancreatic lipase (PPL) activity. Sitosterol is an important compound extracted from the leaves of Aloe vera . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.