Learn More
Medchemexpress LLC Beta-sitosterol | 83-46-5 | MFCD00003631 | 98.2% | 414.71 g/mol | C29H50O | 10 MG
SDP
Catalog No. 5000296634
Encompass_Preferred
Beta-sitosterol is a plant-derived sterol supplied as a 10 mg solid analytical reference standard intended for research and analytical applications, including method development, assay calibration, identification, and purity assessment. Reported attributes: purity ~98.18%, CAS 83-46-5, molecular formula C29H50O, molecular weight 414.71 g/mol.
  • Solid analytical reference standard.
  • Single 10 mg quantity.
  • Reported purity 98.2%.
  • CAS number 83-46-5.
  • Suitable for method development and assay calibration.
  • Store and handle according to SDS instructions.
Catalog No. 50-002-96634 Supplier Medchemexpress LLC Supplier No. HYN0171R10MG
Beta-sitosterol is a plant-derived sterol supplied as a 10 mg solid analytical reference standard intended for research and analytical applications, including method development, assay calibration, identification, and purity assessment. Reported attributes: purity ~98.18%, CAS 83-46-5, molecular formula C29H50O, molecular weight 414.71 g/mol.
  • Solid analytical reference standard.
  • Single 10 mg quantity.
  • Reported purity 98.2%.
  • CAS number 83-46-5.
  • Suitable for method development and assay calibration.
  • Store and handle according to SDS instructions.