Learn More
Abcam beta-Sitosterol, Antitumor agent, 5G
Catalog No. 502706132
Encompass_Preferred
MW 414.7 Da, Purity >98%. Plant sterol. Various pharmacological effects. Antitumor agent. Activates caspase 3 and induces G2/M arrest and DNA fragmentation. Increases α-tubulin levels and promotes microtubule polymerization. Inhibits dietary cholesterol absorption in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6132 Supplier Abcam Supplier No. AB1431225G
Add to Cart
Add to Cart
MW 414.7 Da, Purity >98%. Plant sterol. Various pharmacological effects. Antitumor agent. Activates caspase 3 and induces G2/M arrest and DNA fragmentation. Increases α-tubulin levels and promotes microtubule polymerization. Inhibits dietary cholesterol absorption in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement