Learn More
Medchemexpress LLC Beta-sitosterol-d7 (mixture of diastereomers) | 2260669-28-9 | MFCD08704917 | ≥98.0% | C29H43D7O | 10MG
SDP
Catalog No. 5000260607
Encompass_Preferred
β-Sitosterol-d7 (mixture of diastereomers) is the deuterium-labeled analogue of β-sitosterol provided as a reference material and internal standard for lipid analysis and mass spectrometry. The material contains seven deuterium atoms and is supplied as a white to off-white solid with high isotopic enrichment suitable for GC-MS and LC-MS quantification and method development.
  • Deuterium-labeled internal standard for accurate quantification.
  • Mixture of diastereomers matching reference analyte behavior.
  • High isotopic enrichment (d7) for clear mass separation.
  • Suitable for GC-MS and LC-MS method development and validation.
  • White to off-white solid, stable under typical storage conditions.
Catalog No. 50-002-60607 Supplier Medchemexpress LLC Supplier No. HY132382S10MG
β-Sitosterol-d7 (mixture of diastereomers) is the deuterium-labeled analogue of β-sitosterol provided as a reference material and internal standard for lipid analysis and mass spectrometry. The material contains seven deuterium atoms and is supplied as a white to off-white solid with high isotopic enrichment suitable for GC-MS and LC-MS quantification and method development.
  • Deuterium-labeled internal standard for accurate quantification.
  • Mixture of diastereomers matching reference analyte behavior.
  • High isotopic enrichment (d7) for clear mass separation.
  • Suitable for GC-MS and LC-MS method development and validation.
  • White to off-white solid, stable under typical storage conditions.