Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Sitosterol From Soybeans Practical MP Biomedicals

Catalog No. icn10288610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10288610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10288610 Supplier MP Biomedicals, Inc Supplier No. 0210288610
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
Molecular Weight (g/mol) 414.718
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C

Specifications

Quantity 10 g
Formula Weight 414.7
Grade Practical
Chemical Name or Material Beta-Sitosterol, From Soybeans
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.