Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Beta Storage Box
SCI_ED

Catalog No. s29136 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
18 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
S29136 18 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S29136
This item is not returnable. View return policy

Holds two tube holders side-by-side

  • Safely stores reagent bottles containing beta-emitting isotopes
  • Made of 0.375 in. thick (0.95 cm) clear acrylic with nonskid feet
  • Cover has handle and removes easily
  • Dimensions: 9L x 6W x 7 in.H (23 x 15 x 18 cm)

Specifications

Includes Lid and radiation warning labels, non-skid feet
Length (Metric) 23 cm
Color Clear
Dimensions (L x W x H) 9 x 6 x 7 in. (23 x 15 x 18 cm)
For Use With (Application) To store various racks, reagent bottles, and other vessels containing beta-emitting isotopes
Material Acrylic
Height (English) 7 in.
Height (Metric) 18 cm
Product Type Beta storage box
Length (English) 9 in.
Thickness (English) 0.37 in.
Holds 2 Tubes side-by-side
Width (English) 6 in.
Width (Metric) 15 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.