Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta Tubulin Antibody | 50 kDa | 50 UL
SDP

Catalog No. 5000396492
Encompass_Preferred

This beta Tubulin Antibody is provided as a liquid at 1 mg/mL, reacting with human, mouse, and rat samples. It targets a protein with a predicted band size of 50 kDa and is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, and Flow Cytometry.

  • Rabbit host, YA585 clonality
  • SwissProt IDs: P07437 (Human), P99024 (Mouse), P69897 (Rat)
  • Liquid formulation, non-conjugated
  • Concentration: 1 mg/mL
  • Applications: WB, ICC/IF, IHC-P, FC
  • Dilution ratios: WB: 1:5000-1:10,000; ICC: 1:50-1:200; IHC-P: 1:100-1:400; FC: 1:100
  • Formulated in 1xTBS (pH 7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Storage: -20°C for 1 year; avoid freeze/thaw cycles
  • Shipping: Blue ice

Catalog No. 50-003-96492 Supplier Medchemexpress LLC Supplier No. HYP8048750UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This beta Tubulin Antibody is provided as a liquid at 1 mg/mL, reacting with human, mouse, and rat samples. It targets a protein with a predicted band size of 50 kDa and is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, and Flow Cytometry.

  • Rabbit host, YA585 clonality
  • SwissProt IDs: P07437 (Human), P99024 (Mouse), P69897 (Rat)
  • Liquid formulation, non-conjugated
  • Concentration: 1 mg/mL
  • Applications: WB, ICC/IF, IHC-P, FC
  • Dilution ratios: WB: 1:5000-1:10,000; ICC: 1:50-1:200; IHC-P: 1:100-1:400; FC: 1:100
  • Formulated in 1xTBS (pH 7.4), 0.05% BSA, 40% Glycerol, and 0.05% Sodium Azide
  • Storage: -20°C for 1 year; avoid freeze/thaw cycles
  • Shipping: Blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.