Learn More
Medchemexpress LLC Beta Tubulin Antibody (YA585) | 55 kDa | 100 UL
SDP
Catalog No. 5000395996
Encompass_Preferred
Beta Tubulin Antibody (YA585) is a non-conjugated, Rabbit-originated IgG monoclonal antibody that targets beta Tubulin, commonly used as a loading control antibody. This liquid antibody has a concentration of 0.2 mg/mL and is reactive with Human, Mouse, and Rat samples. It should be stored at -20°C for up to 1 year, avoiding repeated freeze/thaw cycles.
  • Verified for Western Blot (WB)
  • Verified for Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Verified for Immunohistochemistry-Paraffin (IHC-P)
  • Verified for Flow Cytometry (FC)
  • Reactivity with Human, Mouse, and Rat samples
  • Rabbit host
  • Monoclonal clonality
  • Non-conjugated
  • Targets Beta Tubulin
Catalog No. 50-003-95996 Supplier Medchemexpress LLC Supplier No. HYP80487100UL
Beta Tubulin Antibody (YA585) is a non-conjugated, Rabbit-originated IgG monoclonal antibody that targets beta Tubulin, commonly used as a loading control antibody. This liquid antibody has a concentration of 0.2 mg/mL and is reactive with Human, Mouse, and Rat samples. It should be stored at -20°C for up to 1 year, avoiding repeated freeze/thaw cycles.
  • Verified for Western Blot (WB)
  • Verified for Immunocytochemistry/Immunofluorescence (ICC/IF)
  • Verified for Immunohistochemistry-Paraffin (IHC-P)
  • Verified for Flow Cytometry (FC)
  • Reactivity with Human, Mouse, and Rat samples
  • Rabbit host
  • Monoclonal clonality
  • Non-conjugated
  • Targets Beta Tubulin