Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta tubulin antibody (YA839) | 10 UL
SDP

Catalog No. 5000442308
Encompass_Preferred

The Beta Tubulin Antibody (YA839) is a mouse-hosted antibody from MedChemExpress, supplied as a liquid at a concentration of 2 mg/mL. It is suitable for laboratory research applications such as Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF), reacting across multiple species including human, mouse, and monkey.

  • Host: Mouse
  • Reactivity: Human, mouse, monkey, goat, hamster, rat, Chlamydomonas Reinhardtii
  • Observed molecular weight: 55 kDa
  • Formulation: PBS with 50% glycerol, 0.5% BSA, 0.02% sodium azide (pH 7.3)
  • Storage: -20°C for up to 1 year; avoid freeze/thaw cycles
  • Applications: WB (1:5000-1:10000), ICC/IF (1:100-1:200)

Catalog No. 50-004-42308 Supplier Medchemexpress LLC Supplier No. HYP8095510UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Beta Tubulin Antibody (YA839) is a mouse-hosted antibody from MedChemExpress, supplied as a liquid at a concentration of 2 mg/mL. It is suitable for laboratory research applications such as Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF), reacting across multiple species including human, mouse, and monkey.

  • Host: Mouse
  • Reactivity: Human, mouse, monkey, goat, hamster, rat, Chlamydomonas Reinhardtii
  • Observed molecular weight: 55 kDa
  • Formulation: PBS with 50% glycerol, 0.5% BSA, 0.02% sodium azide (pH 7.3)
  • Storage: -20°C for up to 1 year; avoid freeze/thaw cycles
  • Applications: WB (1:5000-1:10000), ICC/IF (1:100-1:200)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.