Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta Tubulin Antibody (YA839) | 100 UL
SDP

Catalog No. 5000441877
Encompass_Preferred

The Beta Tubulin Antibody (YA839) is a non-conjugated liquid supplied at a concentration of 2 mg/mL. Formulated in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, this antibody is suitable for laboratory research and the manufacture of substances. It exhibits reactivity with samples from human, mouse, monkey, goat, hamster, rat, and Chlamydomonas Reinhardtii.

  • Reactivity: Human, mouse, monkey, goat, hamster, rat, Chlamydomonas Reinhardtii
  • Observed molecular weight: 55 kDa
  • Applications: WB, ICC/IF
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Formulation: PBS with 50% glycerol, 0.5% BSA, and 0.02% sodium azide

Catalog No. 50-004-41877 Supplier Medchemexpress LLC Supplier No. HYP80955100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Beta Tubulin Antibody (YA839) is a non-conjugated liquid supplied at a concentration of 2 mg/mL. Formulated in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, this antibody is suitable for laboratory research and the manufacture of substances. It exhibits reactivity with samples from human, mouse, monkey, goat, hamster, rat, and Chlamydomonas Reinhardtii.

  • Reactivity: Human, mouse, monkey, goat, hamster, rat, Chlamydomonas Reinhardtii
  • Observed molecular weight: 55 kDa
  • Applications: WB, ICC/IF
  • Storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Formulation: PBS with 50% glycerol, 0.5% BSA, and 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.