Learn More
Santa Cruz Biotechnology BETA TUBULIN D-10
Supplier:  Santa Cruz Biotechnology SC5274
Encompass
beta Tubulin (D-10), 200microgram/mL, Isotope: mouse IgG, Epitope: 210-444 (human), Application: WB, IP and IHC(P), Species: Mouse, Rat, Human, MW: 55 kd, raised against a recombinant protein, Storage Temperature: 4degreeC
Catalog No. NC9792937