Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-Zearalanol | 42422-68-4 | 5 MG
SDP

Catalog No. 5000417749
Encompass_Preferred

Beta-Zearalenol is a mycotoxin produced by Fusarium spp, which induces apoptosis and oxidative stress in mammalian reproductive cells. It is a derivative of zearalenone (ZEA) and can conjugate with glucuronic acid.

  • Purity 99.83%
  • Appearance solid, white to off-white
  • Molecular weight 322.40
  • Formula C18H26O5
  • Source microorganisms
  • Inhibits BGC proliferation in a dose-dependent manner
  • Shows high toxicity in HepG2 cells
  • Exhibits synergistic effects with ZEA or α-ZOL in HepG2 cells
  • Highest inhibitory effect on IL-8, IL-1β, and TNF-α

Catalog No. 50-004-17749 Supplier Medchemexpress LLC Supplier No. HYN67405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta-Zearalenol is a mycotoxin produced by Fusarium spp, which induces apoptosis and oxidative stress in mammalian reproductive cells. It is a derivative of zearalenone (ZEA) and can conjugate with glucuronic acid.

  • Purity 99.83%
  • Appearance solid, white to off-white
  • Molecular weight 322.40
  • Formula C18H26O5
  • Source microorganisms
  • Inhibits BGC proliferation in a dose-dependent manner
  • Shows high toxicity in HepG2 cells
  • Exhibits synergistic effects with ZEA or α-ZOL in HepG2 cells
  • Highest inhibitory effect on IL-8, IL-1β, and TNF-α

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.