Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich beta-Zearalenol

Catalog No. Z20005MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z20005MG 5 mg
Z200010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z20005MG Supplier Sigma Aldrich Supplier No. Z20005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant involved in biological studies including:
  • Synthesis of amino glycoside nucleotides
  • Enzymatic synthesis of glucuronides of zearalenone
  • Derivatization of zearalenone

Specifications

CAS 71030-11-0
Molecular Formula C18H24O5
Linear Formula C18H24O5
MDL Number MFCD00151078
Synonym beta-Zearalenol; 2,4-Dihydroxy-6-(6 beta,10-dihydroxy-trans-1-undecenyl)benzoic acid mu-lactone
Molecular Weight (g/mol) 320.38
Quantity 5 mg
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.