Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque beta 2-Microglobulin / B2M Protein;HEK293;100UG;B2M,B2M,Beta-2 microglobulin
Supplier:  ACROBiosystems B2MC5226100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque beta 2-Microglobulin / B2M Protein;100UG;Cynomolgus / Rhesus macaque B2M, His Tag (B2M-C5226) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession # Q8SPW0).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.6 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-9126