Learn More
ACROBiosystems Beta-2-Microglobulin, Cynomolgus and Rhesus Macaque, His Tag, 1 mg
Catalog No. 501659127
Encompass_Preferred
Beta-2-microglobulin (B2M) is a component of MHC class I molecules and belongs to the beta-2-microglobulin family. Expressed in human 293 cells, this recombinant protein is supplied as a lyophilized powder. It contains a polyhistidine tag at the C-terminus and is designed for applications requiring high-purity B2M.
  • Purity is >95% as determined by SDS-PAGE
  • Calculated molecular weight is 12.6 kDa, migrating as 14 kDa under reducing conditions
  • Endotoxin level is less than 1.0 EU per μg by the LAL method
  • Formulated in PBS, pH 7.4, with trehalose as a protectant
  • Stable for 12 months in lyophilized state when stored at -20°C to -70°C
  • Avoid repeated freeze-thaw cycles
Catalog No. 50-165-9127 Supplier ACROBiosystems Supplier No. B2MC52261MG
Beta-2-microglobulin (B2M) is a component of MHC class I molecules and belongs to the beta-2-microglobulin family. Expressed in human 293 cells, this recombinant protein is supplied as a lyophilized powder. It contains a polyhistidine tag at the C-terminus and is designed for applications requiring high-purity B2M.
  • Purity is >95% as determined by SDS-PAGE
  • Calculated molecular weight is 12.6 kDa, migrating as 14 kDa under reducing conditions
  • Endotoxin level is less than 1.0 EU per μg by the LAL method
  • Formulated in PBS, pH 7.4, with trehalose as a protectant
  • Stable for 12 months in lyophilized state when stored at -20°C to -70°C
  • Avoid repeated freeze-thaw cycles