Learn More
Sino Biological Recombinant Anti-beta-Catenin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11279R021100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human beta-Catenin / CTNNB1 (rh beta-Catenin / CTNNB1; Catalog#11279-H20B; P35222-1; Met1-Leu781).
Catalog No. 50-228-3211