Learn More
Medchemexpress LLC Betacellulin/BTC Cy 50ug | 50ug
SDP
Catalog No. 5000186627
Encompass_Preferred
Betacellulin/BTC Protein a member of the epidermal growth factor (EGF) family is known to play an important role in regulating growth and differentiation of pancreatic beta cells And it is a growth factor that binds to EGFR ERBB4 and other EGF receptor family members In addition BTC exerts potent mitogenic effects on retinal pigment epithelial cells and vascular smooth muscle cells as a monomer Betacellulin/BTC Protein Cynomolgus (HEK293 Fc) is the recombinant cynomolgus-derived Betacellulin/BTC protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-86627 Supplier Medchemexpress LLC Supplier No. HYP7730350UG
Betacellulin/BTC Protein a member of the epidermal growth factor (EGF) family is known to play an important role in regulating growth and differentiation of pancreatic beta cells And it is a growth factor that binds to EGFR ERBB4 and other EGF receptor family members In addition BTC exerts potent mitogenic effects on retinal pigment epithelial cells and vascular smooth muscle cells as a monomer Betacellulin/BTC Protein Cynomolgus (HEK293 Fc) is the recombinant cynomolgus-derived Betacellulin/BTC protein expressed by HEK293 with C-hFc labeled tag