Learn More
Medchemexpress LLC Betacellulin/BTC Mo 100ug
SDP
Supplier:  Medchemexpress LLC HYP7329100UG
Encompass_Preferred
Probetacellulin Protein Mouse induces differentiation of pancreatic -cells and promotes regeneration of -cells in experimental diabetes
Catalog No. 50-001-81315