Learn More
Medchemexpress LLC Betacellulin/BTC Mo 10ug

Supplier: Medchemexpress LLC HYP714210UG
Betacellulin (BTC) Protein, mouse, is a recombinant growth factor produced in HEK293 cells and supplied as a lyophilized powder for research use. It contains the bioactive EGF-like domain (sequence range D32-Y111) and is provided with specifications for activity, purity, and storage to support in vitro assays and cell-based studies.
- Recombinant mouse protein expressed in HEK293 cells.
- Lyophilized powder formulation for convenient handling.
- Biological activity: ED50 <0.08 ng/mL (3T3 cell assay).
- Low endotoxin: <0.2 EU/μg, determined by LAL method.
- Includes defined amino acid sequence corresponding to the active domain.
- Formulated from 0.22 μm filtered PBS and lyophilized for stability.
- Storage: store at -20°C; after reconstitution, 4°C for 1 week or -20°C (with carrier) for longer.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.