Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0316250
Encompass
Betacellulin (BTC) is a member of the EGF family of growth factors that also includes EGF TGF-Alpha Amphiregulin HB-EGF Epiregulin Tomoregulin Heregulin and Neuregulins. Mature human BTC protein exhibits 80% amino acidsimilarity with mouse BTC protein. BTC is expressed in most tissues including kidney uterus liver and pancreas. It is also present in body fluids including serum milk and colostrum. It is synthesized primarily as a transmembrane precursor which is then processed to a mature molecule by proteolytic events. BTC signals through the EGF receptor.
Catalog No. 50-252-9762