Learn More
ATCC Betacoronavirus 1 RNA; Strain: OC43; Human coronavirus OC43
Catalog No. 502381718
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from HCT-8 cells (ATCC CCL-244) infected with Betacoronavirus 1 strain OC43 (ATCC VR-1558).This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-1718 Supplier ATCC Supplier No. VR1558D
Genomic RNA isolated from a preparation of cell lysate and supernatant from HCT-8 cells (ATCC CCL-244) infected with Betacoronavirus 1 strain OC43 (ATCC VR-1558).This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.