Learn More
ATCC Betacoronavirus 1 strain OC43 (ATCC VR-1558™)
Catalog No. 502382002
Encompass
Betacoronavirus 1 strain OC43 (ATCC VR-1558™) is a whole-genome sequenced virus used in respiratory disease research and molecular assay development. Originally isolated from a patient with cold-like symptoms, it is propagated in HCT-8 cells and stored frozen.
Recommended companion products:
HCT-8 cells (Fisher Cat. No. 50-238-3076, ATCC CCL-244™)
RPMI-1640 medium (Fisher Cat No. 50-238-3277, ATCC 30-2001™).
Catalog No. 50-238-2002 Supplier ATCC Supplier No. VR1558
Betacoronavirus 1 strain OC43 (ATCC VR-1558™) is a whole-genome sequenced virus used in respiratory disease research and molecular assay development. Originally isolated from a patient with cold-like symptoms, it is propagated in HCT-8 cells and stored frozen.
Recommended companion products:
HCT-8 cells (Fisher Cat. No. 50-238-3076, ATCC CCL-244™)
RPMI-1640 medium (Fisher Cat No. 50-238-3277, ATCC 30-2001™).