Learn More
Biomatik Corporation Beta-Crosslaps (Bctx) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31287
Encompass_Preferred
Beta-Crosslaps (Bctx) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Beta-Crosslaps (bCTx). The Beta-Crosslaps (Bctx) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Applications: WB, IHC, ICC, IP. Target Synonyms: Beta-Crosslaps (bCTx). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6553