Learn More
Med Vet International Betadine Surgical Scrub 7.5% Veterinary Gallon, 4 Count
Catalog No. 503045197
Encompass_Preferred
A sudsing cleanser that contains 7.5% povidone-iodine. Povidone-iodine, the active ingredient in Betadine products, works against a wide variety of pathogens in vitro. Helps to reduce bacteria on the skin that can potentially cause infection. For preparation of the skin prior to surgery For handwashing to reduce bacteria on the skin Significantly reduces the number of microorganisms on the hands and forearms prior to surgery or patient care
Catalog No. 50-304-5197 Supplier Med Vet International Supplier No. OTCBVET1G
A sudsing cleanser that contains 7.5% povidone-iodine. Povidone-iodine, the active ingredient in Betadine products, works against a wide variety of pathogens in vitro. Helps to reduce bacteria on the skin that can potentially cause infection. For preparation of the skin prior to surgery For handwashing to reduce bacteria on the skin Significantly reduces the number of microorganisms on the hands and forearms prior to surgery or patient care