Learn More
Med Vet International Betadine Surgical Scrub, 1 Gallon
Catalog No. 502828828
Encompass_Preferred
Betadine Scrub is an antiseptic, microbicidal, sudsing skin cleanser for pre-operative scrubbing and washing by surgeons and pre-operative preparation of patients' skin. Contains 7.5% povidone iodine (0.75% available iodine). 100% effective and non-irritating.
Catalog No. 50-282-8828 Supplier Med Vet International Supplier No. PHVBETASCRUB1G
Betadine Scrub is an antiseptic, microbicidal, sudsing skin cleanser for pre-operative scrubbing and washing by surgeons and pre-operative preparation of patients' skin. Contains 7.5% povidone iodine (0.75% available iodine). 100% effective and non-irritating.