Learn More
Med Vet International Betagen Topical Spray, Broad Spectrum Antibiotic, 60mL
Supplier:  Med Vet International RXGENTOP
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
For topical use in dogs only. Gentamicin is a broad-spectrum antibiotic. It is a highly effective topical treatment for bacterial infections of the skin. Betamethasone valerate, is a synthetic glucocorticoid. In veterinary medicine, betamethasone has been shown to provide anti-inflammatory and antipruritic activity in the topical management of corticosteroid-responsive infected superficial lesions in dogs. 60mL per bottle.
Catalog No. 50-282-8156