Learn More
Cayman Chemical Betahistine-d3 (hydrochloride), Purity: ≥99% deuterated forms (d1-d3), CAS: 244094-72-2, 10 mg
SDP
Catalog No. 502909695
Encompass_Preferred
An internal standard for the quantification of betahistine by GC- or LC-MS
Catalog No. 50-290-9695 Supplier Cayman Chemical Supplier No. 3555910 MG
An internal standard for the quantification of betahistine by GC- or LC-MS