Learn More
Selleck Chemical LLC Betahistine 2HCl S3176-50mg
SDP
Catalog No. 503109860
Encompass_Preferred
Betahistine (PT-9) is a histamine H3 receptor inhibitor with IC50 of 1 9 M
Catalog No. 50-310-9860 Supplier Selleck Chemical LLC Supplier No. S317650MG
Betahistine (PT-9) is a histamine H3 receptor inhibitor with IC50 of 1 9 M