Learn More
Medchemexpress LLC Betahistine-d3 dihydrochloride | 244094-72-2 | 99.0% | 5 MG
SDP
Catalog No. 5000408552
Encompass_Preferred
Betahistine-d3 dihydrochloride is a deuterium-labeled form of Betahistine dihydrochloride, an orally active histamine H1 receptor agonist and H3 receptor antagonist used in the study of rheumatoid arthritis (RA). Stable heavy isotopes, like deuterium in this molecule, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process, and deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08552 Supplier Medchemexpress LLC Supplier No. HYB0524AS5MG
Betahistine-d3 dihydrochloride is a deuterium-labeled form of Betahistine dihydrochloride, an orally active histamine H1 receptor agonist and H3 receptor antagonist used in the study of rheumatoid arthritis (RA). Stable heavy isotopes, like deuterium in this molecule, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process, and deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.