Learn More
Medchemexpress LLC Betahistine dihydrochloride | 5579-84-0 | 99.8% | 5 G
SDP
Catalog No. 5000407571
Encompass_Preferred
Betahistine dihydrochloride is an orally active histamine H1 receptor agonist and a H3 receptor antagonist. It is commonly used in the study of rheumatoid arthritis (RA).
  • Orally active histamine H1 receptor agonist
  • H3 receptor antagonist
  • Used for the study of rheumatoid arthritis (RA)
  • Inhibits [125I]iodoproxyfan binding to CHO cell membranes
  • Regulates cAMP formation in CHO cells
  • Attenuates severity of arthritis and reduces pro-inflammatory cytokines in paw tissues of CIA mice
  • Ameliorates mouse CIA by decreasing joint destruction
Catalog No. 50-004-07571 Supplier Medchemexpress LLC Supplier No. HYB0524A5G
Betahistine dihydrochloride is an orally active histamine H1 receptor agonist and a H3 receptor antagonist. It is commonly used in the study of rheumatoid arthritis (RA).
  • Orally active histamine H1 receptor agonist
  • H3 receptor antagonist
  • Used for the study of rheumatoid arthritis (RA)
  • Inhibits [125I]iodoproxyfan binding to CHO cell membranes
  • Regulates cAMP formation in CHO cells
  • Attenuates severity of arthritis and reduces pro-inflammatory cytokines in paw tissues of CIA mice
  • Ameliorates mouse CIA by decreasing joint destruction