Learn More
Medchemexpress LLC Betahistine dihydrochloride | 5579-84-0 | 99.8% | 500 MG
SDP
Catalog No. 5000408306
Encompass_Preferred
Betahistine dihydrochloride is an orally active histamine H1 receptor agonist and H3 receptor antagonist. It is used for the study of rheumatoid arthritis (RA).
  • Orally active histamine H1 receptor agonist
  • Histamine H3 receptor antagonist
  • Inhibits [125I]iodoproxyfan binding to CHO cell membranes
  • Regulates cAMP formation in CHO cells
  • Increases tele-methylhistamine (t-MeHA) levels in vivo
  • Attenuates the severity of arthritis in CIA mice
  • Reduces pro-inflammatory cytokines in paw tissues
Catalog No. 50-004-08306 Supplier Medchemexpress LLC Supplier No. HYB0524500MG
Betahistine dihydrochloride is an orally active histamine H1 receptor agonist and H3 receptor antagonist. It is used for the study of rheumatoid arthritis (RA).
  • Orally active histamine H1 receptor agonist
  • Histamine H3 receptor antagonist
  • Inhibits [125I]iodoproxyfan binding to CHO cell membranes
  • Regulates cAMP formation in CHO cells
  • Increases tele-methylhistamine (t-MeHA) levels in vivo
  • Attenuates the severity of arthritis in CIA mice
  • Reduces pro-inflammatory cytokines in paw tissues