Learn More
Medchemexpress LLC HY-D0237 100mg Medchemexpress, Betahistine (mesylate) CAS:54856-23-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0237100MG
Encompass_Preferred
Medchemexpress, HY-D0237 100mg Betahistine (mesylate) CAS:54856-23-4 Betahistine mesylate is an orally active histamine H1 receptor agonist and a H3 receptor antagonist. Betahistine mesylate is used for the study of rheumatoid arthritis (RA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2746