Learn More
Cayman Chemical Betahistine (hydrochloride), Purity: ≥98%, CAS: 5579-84-0, 1 g
SDP
Catalog No. 502940252
Encompass_Preferred
A histamine H3 receptor antagonist and histamine H1 receptor agonist; dilates pre-capillary arterioles and increases blood flow to the stria vascularis in guinea pigs; reduces olanzapine-induced weight gain and increased feeding behavior in female rats at 8 mg/kg
Catalog No. 50-294-0252 Supplier Cayman Chemical Supplier No. 276021 G
A histamine H3 receptor antagonist and histamine H1 receptor agonist; dilates pre-capillary arterioles and increases blood flow to the stria vascularis in guinea pigs; reduces olanzapine-induced weight gain and increased feeding behavior in female rats at 8 mg/kg