Learn More
Cayman Chemical Betaine Aldehyde (chloride), Purity: ≥95%, CAS: 7758-31-8, 50 mg
SDP
Catalog No. 502965786
Encompass_Preferred
The physiological intermediate in the oxidation of choline to betaine
Catalog No. 50-296-5786 Supplier Cayman Chemical Supplier No. 1727050 MG
The physiological intermediate in the oxidation of choline to betaine